5-Bromo-1,2,3-trimethoxybenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

B4739-25G B4739-5G
TCB4739-25GEA 263.12 CAD
TCB4739-25G TCB4739-5G
5-Bromo-1,2,3-trimethoxybenzene ≥98.0% (by GC)
5-Bromo-1,2,3-trimethoxybenzene
Formula: C₉H₁₁BrO₃
Boiling Pt: 273°C
MDL Number: MFCD00017169
CAS Number: 2675-79-8
EINECS: 220-223-9

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 78.0 to 82.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR