4,5-Dibromoveratrole ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
D5019-25G
D5019-5G
76219-780EA
244.9
CAD
76219-780
76219-782
4,5-Dibromoveratrole ≥98.0% (by GC)
4,5-Dibromoveratrole
Formula:
C₈H₈Br₂O₂ |
MDL Number:
MFCD00014894 CAS Number: 37895-73-1 EINECS: 253-705-2 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 87.0 to 91.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...