1-Bromo-2-chloro-4-nitrobenzene ≥97.0% (by GC)

Supplier: TCI America
Warning

B5348-25G B5348-5G
76219-582EA 139.04 CAD
76219-582 76219-584
1-Bromo-2-chloro-4-nitrobenzene ≥97.0% (by GC)
1-Bromo-2-chloro-4-nitrobenzene
Formula: C₆H₃BrClNO₂
MDL Number: MFCD00051514
CAS Number: 29682-39-1
EINECS: 249-773-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 59.0 to 63.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR