9-Nitroanthracene ≥90.0% (by GC)

Supplier: TCI America

N0987-1G N0987-5G
TCN0987-1GEA 82.16 CAD
TCN0987-1G TCN0987-5G
9-Nitroanthracene ≥90.0% (by GC)
9-Nitroanthracene
Formula: C₁₄H₉NO₂
Boiling Pt: 275°C
MDL Number: MFCD00001248
CAS Number: 602-60-8
EINECS: 210-021-9

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 90.0 %
Melting point 142.0 to 147.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR