3-Methyl-1-butene ∼12.5% ∼1.5 mol/L in THF

Supplier: TCI America
Danger

M2565-100ML M2565-500ML
TCM2565-100MLEA 114.08 CAD
TCM2565-100ML TCM2565-500ML
3-Methyl-1-butene ∼12.5% ∼1.5 mol/L in THF
3-Methyl-1-butene
Formula: C₅H₁₀
Storage Temperature: Refrigerator
MDL Number: MFCD00008937
CAS Number: 563-45-1
EINECS: 209-249-1
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Concentration (HNMR) 12.0 to 14.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR