3-Methyl-1-butene ∼15% ∼2.5 mol/L in dichloromethane

Supplier: TCI America
Danger

M2563-100ML M2563-500ML
TCM2563-100MLEA 140.46 CAD
TCM2563-100ML TCM2563-500ML
3-Methyl-1-butene ∼15% ∼2.5 mol/L in dichloromethane
3-Methyl-1-butene
Formula: C₅H₁₀
Storage Temperature: Refrigerator
MDL Number: MFCD00008937
CAS Number: 563-45-1
EINECS: 209-249-1
UN: 1593
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Concentration (HNMR) 15.0% to 17.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR