2-Methylpropene ∼10% in isopropyl ether

Supplier: TCI America
Danger

I0910-100ML I0910-500ML
TCI0910-100MLEA 86.82 CAD
TCI0910-100ML TCI0910-500ML
2-Methylpropene ∼10% in isopropyl ether
2-Methylpropene
Formula: C₄H₈
Storage Temperature: Refrigerator
MDL Number: MFCD00008898
CAS Number: 115-11-7
EINECS: 204-066-3
UN: 1993
ADR: 3,II
Merck Index: 14,05141

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Concentration (HNMR) 10.0 to 12.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR