Tridodecylamine (hydrogen ionophore I) 85,0 GC_ASSAY_METHOD

Supplier: TCI America
Warning

Synonyms: Trilaurylamine, Tridodecylamine

T3519-100ML T3519-25ML
TCT3519-100MLEA 298.62 CAD
TCT3519-100ML TCT3519-25ML
Tridodecylamine (hydrogen ionophore I) 85,0 GC_ASSAY_METHOD
Tridodecylamine (Hydrogen ionophore I)
Formula: C₃₆H₇₅N
MW: 522 g/mol
Boiling Pt: 450
Melting Pt: 16
Flash Pt: 190°C
MDL Number: MFCD00008971
CAS Number: 102-87-4
EINECS: 203-063-4

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid to cloudy liquid
Purity (GC) min. 85.0 %
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR