1-Chloro-6-iodohexane ≥98.0% (by GC) stabilized
Supplier: TCI America
|
Warning
|
Stabilization:
Stabilized with Copper chip
C2714-25G
C2714-5G
76219-680EA
344.44
CAD
76219-680
76219-682
1-Chloro-6-iodohexane ≥98.0% (by GC) stabilized
1-Chloro-6-iodohexane
Formula:
C₆H₁₂ClI Boiling Pt: 92°C |
MDL Number:
MFCD00039417 CAS Number: 34683-73-3 EINECS: 252-144-0 |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...