SafeMask® Master Series Procedure Earloop Face Masks
Supplier: Medicom
2051 2052 2053 2058 2059
77610-660BX 19.42 CAD
77610-660 77610-662 77610-664 77610-674 77610-676
SafeMask® Master Series Procedure Earloop Face Masks
Masks Surgical Masks
Procedure and surgical masks are intended to be worn by personnel during surgical or other medical procedures to protect both the patient and the operating personnel from the transfer of microorganisms, body fluids, and particulate material.
  • Masks are designed in a variety of unique and vibrant colors
  • Engineered with Simply Soft Technology™ for a smooth, luxurious, and comfortable fit
  • Ideal blend of breathability and softness
  • Latex-free
  • Made in USA
A beautiful line of masks designed in a variety of unique and vibrant colors, so that each mask is as personalized as the care provided. Master Series ASTM masks are engineered with Simply Soft Technology™ for a smooth, luxurious, and comfortable fit, offering a perfect blend of breathability and softness.
Masks are constructed from a combination of spunbond Polypropylene (SBPP), melt blown Polypropylene (MBPP), Aluminum or Polypropylene (PP)-covered Iron, knitted Polyester and Spandex or knitted Polypropylene and Spandex.
Certifications: ASTM F 2100 Level 1, Level 2, and Level 3 barrier protection.
Delivery information: Store in cool, clean, and dry conditions. Avoid excessive heat (over 40 °C or 104 °F).
Packaging: Masks are packaged 50 per box, 10 boxes per case.