SafeMask® Procedure Earloop Face Masks
Supplier: Medicom
2016 2017
77610-644BX 17.73 CAD
77610-644 77610-646
SafeMask® Procedure Earloop Face Masks
Masks Surgical Masks
Procedure and surgical masks are intended to be worn by personnel during surgical or other medical procedures to protect both the patient and the operating personnel from the transfer of microorganisms, body fluids, and particulate material.
  • Customized comfort and protection
  • Soft bond ultrasonics ensures durable construction, eliminating holes and defects
  • Latex-free
  • Embossed Medicom logo
  • Made in USA
Highest quality filter media for optimal filtration and breathability, meeting ASTM Level 1 performance requirements. Soft, strong, sonically sealed ample length ear loops ensure there is no pulling. They fit comfortably and are attached to the outside of the mask to eliminate irritation. Adjustable, extra-long, enclosed aluminum nose piece forms strong seal for maximum protection and comfort. Non-woven spunbond outer layer material provides maximum protection against fluid penetration, and will not lint, tear, or shred. Non-woven spunbond inner layer is made of white, dye-free, fluid-resistant material that will not lint, tear, or shred.
Masks are constructed from a combination of spunbond Polypropylene (SBPP), melt blown Polypropylene (MBPP), Aluminum or Polypropylene (PP)-covered Iron, knitted Polyester and Spandex or knitted Polypropylene and Spandex.
Certifications: ASTM F 2100 Level 1 barrier protection.
Delivery information: Store in cool, clean, and dry conditions. Avoid excessive heat (over 40 °C or 104 °F).
Packaging: Masks are packaged 50 per box, 10 boxes per case.