1-Bromo-2-(2-methoxyethoxy)ethane ≥90.0% (by GC) stabilised

Supplier: TCI America

Synonyms: 1-Bromo-3,6-dioxaheptane, 2-(2-Methoxyethoxy)ethyl bromide, m-PEG2-Br

Stabilization: Stabilised with sodium carbonate (Na₂CO₃)

B4736-25G B4736-5G
TCB4736-25GEA 282.79 CAD
TCB4736-25G TCB4736-5G
1-Bromo-2-(2-methoxyethoxy)ethane ≥90.0% (by GC) stabilised
1-Bromo-2-(2-methoxyethoxy)ethane
Formula: BrH₂CCH₂OCH₂CH₂OCH₃
MW: 183.05 g/mol
Boiling Pt: 172
Melting Pt: -23
Flash Pt: 68°C
Storage Temperature: Refrigerator
MDL Number: MFCD00000238
CAS Number: 54149-17-6
EINECS: 259-000-6

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR