Ethyl propenyl ether cis- and trans-mixture ≥98.0% (by GC)
Supplier: TCI America
|
Danger
|
E1052-100ML
E1052-25ML
TCE1052-100MLEA
271.76
CAD
TCE1052-100ML
TCE1052-25ML
Ethyl propenyl ether cis- and trans-mixture ≥98.0% (by GC)
Ethyl propenyl ether
Formula:
C₅H₁₀O Boiling Pt: 70°C Flash Pt: 20°C |
MDL Number:
MFCD00015184 CAS Number: 928-55-2 EINECS: 213-176-0 UN: 3271 ADR: 3,II |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % (total of isomers) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...