Marble Balance Table, Gawet
Supplier: Gawet
SLAB A4-1602
12568-006EA 1723.39 CAD
12568-006 12568-004
Marble Balance Table, Gawet
Furniture Tables and Workbenches
The 7.6 cm (3") thick marble slab construction provides maximum damping effect and vibration-free stability for analytical instrument scales.
  • Color: white with brown, green gray markings
Marble slab legs are reinforced with an iron cross-beam coated for corrosion resistance. All edges are rounded and precisely cut to ensure a smooth, level working surface.
Ordering information: Single 5 cm (2") thick marble slabs are available for creating stable work areas on countertops; contact your Avantor sales representative for more information.
Order Now
SPECIFICATIONS
Environmentally Preferable