FARRAR® ULC Ultra-Low Chambers
Supplier: TRANE TECHNOLOGIES LIFE SCIENCES LL
ULC259-DWC60HZ ULC259-SAC60HZ ULC259-SWC60HZ ULC328-SAC60HZ ULC328-SWC60HZ ULC190-SAC60HZ ULC190-SWC60HZ ULC259-BAC60HZ ULC259-BWC60HZ ULC259-DAC60HZ
77592-006EA 388281.68 CAD
77592-006 77592-008 77592-010 77592-012 77592-014 77591-996 77591-998 77592-000 77592-002 77592-004
FARRAR® ULC Ultra-Low Chambers
Freezers Ultra Low Temperature Freezers
Purpose-built for life science applications, FARRAR®’s ULC series including the Ultra-Low Chamber (ULC-190), Ultra-Low Chamber (ULC-328), and the new Ultra-Low Chamber (ULC-259) are the only −20 to −80 °C, forced-air, ultra-low-temperature freezers on the market with air and water-cooled options.
  • Optimized footprint - 50% less floorspace versus standard ULT configurations with six times the capacity (one ULC equals six ULTs)
  • Industry's fastest temperature recovery - 10× faster temperature recovery from door opening helps ensure product integrity
  • Integrated, fully redundant systems (control, refrigerations, airflow) - Eliminates need for backup freezer
  • Automatic defrost - Never worry about taking units out of service to manually defrost
  • Extremely tight temperature uniformity at any setpoint - At +4, −80 °C or any temperature in between, count on better than ±3 °C anywhere inside the ULC, even during a defrost cycle
  • Engineered with material handling in mind - Optional racking, shelving, drawers and carts optimize process and workflow (available for all models)
  • Equipment performance/temperature monitoring (MODbus) - Enables easy connection to nearly every building management and automation system (BAS/BMS)
Forced air convection: An industry game changer - FARRAR® pioneered forced air convection technology in its reach-in chambers. This key differentiator affects uniformity. The chambers uniformly force air, at the targeted temperature, throughout the chamber. This means FARRAR® freezer and freeze/thaw equipment is up to 10 times more efficient at removing heat than the traditional cold-wall heat displacement process. Recovery from door opening is critical to helping ensure product integrity. FARRAR® ULC Series chambers dual refrigeration units recover to −80 °C in less than 25 minutes from a 1 minute door opening, versus several hours experienced with competitor solutions.
Accessories information: Components included in the standard set-ups for each unit are listed below (with supplier part numbers). Please speak to your sales representative about other options and accessories available.
ULC-190 Standard Set-up, Shelves
* ULC 190 Chamber - #6700617
* Refrigeration System - #6700028 (air-cooled), #6000442 (water-cooled)
* Middle Box (Non-Monitoring) - #6000442
* Standard ULC-190 Shelf Kit - #6700645
ULC-259 Standard Set-up, Shelves
* ULC 259 Chamber - #6700619
* Refrigeration System - #6700028 (air-cooled), #600029 (water-cooled)
* Middle Box (Non-Monitoring) - #6000442
* ULC-259 Standard Shelf System - #6700775
ULC-259 Standard Set-up, Drawers
* ULC 259 Chamber - #6700619
* Refrigeration System - #6700028 (air-cooled), #600029 (water-cooled)
* Middle Box (Non-Monitoring) - #6000442
* ULC-259 Drawer System - #6700761
ULC-259 Standard Set-up, Biobank
* ULC 259 Chamber - #6700619
* Refrigeration System - #6700028 (air-cooled), #600029 (water-cooled)
* Middle Box (Non-Monitoring) - #6000442
* ULC-259 Biorack Assembly - #6700769
ULC-328 Standard Set-up, Pallet Rack
* ULC 328 Chamber - #6700615
* Refrigeration System - #6700028 (air-cooled), #600029 (water-cooled)
* Middle Box (Non-Monitoring) - #6000442
* ULC-328 Pallet Rack Assembly - #6700401
Ordering information: Prior to placing an order, please work with your Avantor and Trane Technologies representatives to review various set-up options and determine the best configuration to meet your operational needs.