VWR® Pipette Filler, Synthetic Rubber
Supplier: VWR
VWR®
K200-VWR
53502-205EA 71.43 CAD
53502-205
VWR® Pipette Filler, Synthetic Rubber
Pipettes Pipette Bulbs
Pipette fillers do away with the frequently hazardous practice of drawing liquids into pipettes by mouth.
  • No metal parts to corrode
For use with pipettes from 2 to 50 ml. Made of synthetic rubber with glass ball valves.
Ordering information: Supplied with an adapter for capacities greater than 20 ml.