VWR® Pipette Fillers
Supplier: VWR International
53497-009 53497-053 53497-086 53497-111
53497-009CS 10440.18 CAD
53497-009 53497-053 53497-086 53497-111
VWR® Pipette Fillers
Pipettes Pipette Bulbs
Pipette fillers do away with the frequently hazardous practice of drawing liquids into pipettes by mouth.
  • 100% natural rubber construction
  • For use with 1 to 45 ml pipettes
  • Four color options available
They work safely with toxic, infectious, odoriferous, radioactive, corrosive, or sterile liquids.
Pipettes are made out of 100% natural rubber with three stainless steel ball valves. For use with pipettes of 1 to 45 ml capacity and 4 to 8 mm outside diameter. A volume of 100 ml is attainable with repeated evacuation. With a simple three-step process,More Product Information