Freestanding Gowning Racks, Eagle MHC
Supplier: Eagle MHC
EP2448-CGRR EP2484-CGRR S2460-SGRN S2484-SGRN EP2448-DGRR EP2484-SGRR EP2472-CGRR EP2472-SGRR S2460-DGRR EP2484-DGRR EP2460-DGRR EP2472-DGRN EP2472-DGRR EP2460-DGRN EP2460-SGRN S2448-DGRR S2460-SGRR S2460-CGRR EP2460-SGRR S2472-CGRR S2448-DGRN S2484-SGRR S2484-CGRR S2472-DGRR EP2448-SGRR S2484-DGRR EP2448-DGRN S2448-SGRR S2448-CGRR EP2484-DGRN EP2460-CGRR EP2448-SGRN S2472-SGRR S2460-DGRN EP2472-SGRN EP2484-SGRN
58202-670EA 10120.06 CAD
58202-670 58202-692 58202-150 58202-694 58202-650 58202-138 58202-656 58202-634 58202-136 58202-698 58202-654 58202-638 58202-636 58202-658 58202-142 58202-140 58202-640 58202-960 58202-146 58202-660 58202-144 58202-666 58202-644 58202-688 58202-964 58202-700 58202-686 58202-664 58202-148 58202-962 58202-648 58202-902 58202-704 58202-646 58202-668 58202-966
Freestanding Gowning Racks, Eagle MHC
Furniture Gowning Racks
Hanger bar constructed of heavy gauge type 304 stainless steel with hanger slots spaced on 2 1/2" centers
  • Available in single, double, or cantilevered styles
  • Finishes available are chrome and electropolished finish
  • Units are available for removable or non-removable hangers
All units feature wire shelf with patented QuadTruss construction. Top mat assembly constructed of 10 gauge mat wires welded to full-length 6 gauge inside support wires and serpentine trusses. At front and rear of shelf, mat wires are welded to a four-wire truss assembly constructed of a 2 gauge bottom wire, a 6 gauge serpentine truss wire which creates a bridge-like reinforcement, and two 6 gauge top wires, one directly below mat wires and one directly above, providing a pincer hold on mat wires. Posts are 1" diameter, grooved on 1" increments, and provided with leveling feet. Optional foot plates available to affix gowning rack to the floor.
1" square 3-sided frame along the bottom.
Ordering information: Hangers must be ordered separately; see 58202-168 series.