Samco™ Disposable Transfer Pipettes, Fine Tip, Thermo Scientific
Supplier: Thermo Fisher Scientific
231 231-1S 231-20S 232-11 232-1S 232-20S 233 234 235 252-20S 235-20S 235-1S 234-20S 233-1S 234-1S 233 233-1S 235 235-1S
14670-325CS 1756.97 CAD
14670-325 14670-327 14670-329 14670-330 14670-331 14670-332 14670-333 14670-336 14670-339 73990-052 73990-068 73990-070 73990-072 73990-078 490014-678 490014-726 490014-728 490014-734 490014-736
Samco™ Disposable Transfer Pipettes, Fine Tip, Thermo Scientific
Pipettes Pasteur/Transfer Pipettes
These fine-tip polyethylene transfer pipettes are excellent for the precise dispensing of small drops (45 to 50 per ml) into microbiological Int.Ø.
Strips, ELISA plates, and microcentrifuge and precipitation tubes. Ideal for removing cell layers, filling electrophoresis wells, spotting chromatography plates, and other similar applications.