Strip Shields, Trax
Supplier: Trax Industrial Products
SS40ST37 SS40ST48 SS40ST68 SS40ST89 SS40ST1010
12999-080EA 718.96 CAD
12999-080 12999-082 12999-084 12999-086 12999-088
Strip Shields, Trax
Prefabricated Rooms
Designed for use in applications where access to a clean enclosure is a concern, but traffic patterns are light and restricted to personnel only (no carts or equipment)
Pre-assembled strip shields are 40mil thick and comprise 20.3cm (8") wide material overlapped 5.1cm (2"). Shipping weights vary depending on size.
Ordering information: Dual-Lock™ attachment system is included with each strip shield.