Safety Shower and Eye/Face Washes, Speakman®
Supplier: Speakman
SE-603 SE-623 H.F.O
VF-0603-SEEA 9427.61 CAD
VF-0603-SE VF-0623-SE VF-HFO-SE
Safety Shower and Eye/Face Washes, Speakman®
First Aid Supplies Showers
This combined safety device can be used to flush contaminants from eyes, face, or entire body.
  • Shower head Is 20.3 cm (8") in Ø
  • Impeller action
  • Stay-open valve
  • Ideal where workers are subjected to contaminants
  • Space-saving unit
This space-saving unit consists of a shower and an eyewash with a stainless steel basin.
The shower has a 20.3 cm (8") Ø head with impeller action and stay-open valve. A stainless steel push handle actuates the eye and face wash. Six aerated outlets in each spray head provide a 22.8×20.3 cm (9×8") curtain of aerated water for more extensive washing action.
These washes are ideal for locations where workers are subjected to contaminants.
Certifications: Meets ANSI Z358.1 standards.