Lifesaver® Wall-Mount Emergency Showers, Speakman®
Supplier: Speakman
SE-220 SE-227
VF-0220-SEEA 1214.02 CAD
VF-0220-SE VF-0227-SE
Lifesaver® Wall-Mount Emergency Showers, Speakman®
First Aid Supplies Showers
Reach for the handle of these safety showers for immediate rinsing off of harmful contaminants.
  • For immediate rinsing off of harmful contaminants
  • Impeller action head for more thorough rinsing
  • Stay-open valve does not require operator control
An impeller action head allows more thorough rinsing action. The stay-open valve does not require operator control once activated. Showers feature a 20.3 cm (8") Ø yellow shower head, a 2.5 cm (1") NPTF chrome-plated brass ball valve, and a high visibility powder-coated yellow aluminum pull rod. Stainless steel shower head and/or stainless steel pull rod are available as alternative options.
Certifications: Meet ANSI Z358.1 standard.