Deck-Mounted Gooseneck Mixing Faucets, with Four-Arm Handles, WaterSaver Faucet
Supplier: WaterSaver Faucet
L411 L411-8
59382-042EA 459.34 CAD
59382-042 VFL412-12 VFL412-8 VFT-0340-00
Deck-Mounted Gooseneck Mixing Faucets, with Four-Arm Handles, WaterSaver Faucet
Faucets and Sinks Faucets
Deck-mounted mixing faucets are made of chrome-plated forged brass and provided with a serrated hose end, four-arm handles, and 1cm (3/8") diameter copper tubes on inlets.
Faucets are available with a 15.2 cm (6"), 20.3 cm (8"), or 30.5 cm (12") spread gooseneck. Faucet valves are self-contained renewable units with replaceable stainless steel seats.
Certifications: CSA certified. Comply with ANSI/ASME A112.18.1M.