SP Bel-Art Economy Pipette Pump™ III Pipettors, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37902-0002 F37902-0010 F37902-0025
47751-780EA 153.14 CAD
47751-780 47751-782 47751-784
SP Bel-Art Economy Pipette Pump™ III Pipettors, Bel-Art Products, a part of SP
Pipettes Pipette Controllers
These pipetting devices provide accurate, trouble-free pipetting with simple, one-handed operation.
  • Ergonomic design for exceptional comfort
  • Simple one-handed operation
  • Color-coded for easy identification
  • Thumb wheel for precise aspirating or dispensing
The ergonomic design enhances comfort, utility, and dispensing precision while helping to reduce the risk of injury from repetitive and awkward pipetting motions. The optimally located thumb wheel rotates easily for precision aspirating or dispensing while using a more relaxed grip. The thumb wheel canMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable