VWR® Pipette Pump™ Pipetting Device
Supplier: VWR International
53502-211 53502-222 53502-233 53502-244
53502-211CS 1097.47 CAD
53502-211 53502-222 53502-233 53502-244
VWR® Pipette Pump™ Pipetting Device
Pipettes Pipette Controllers
This sturdy pipetting device accepts both disposable and standard pipettes in the flexible tapered chuck, per ASTM Standard E714-80.
  • Safe, accurate and trouble-free pipetting
  • Simple, one-handed operation
  • Color-coded for quick identification
Press plunger for rapid emptying. Resistant to acids and alkalis, they are color-coded for easy size identification. Disassemble all parts for cleaning and before autoclaving at 121 °C (250 °F) for 20 min except the chuck which should not be autoclaved.
Toothed wheel rotates forward or backward for preciseMore Product Information