Deck-Mount Eyewash/Drench Hoses, WaterSaver Faucet
Supplier: WaterSaver Faucet
EW1022 CTEW1022
59380-032EA 979.69 CAD
59380-032 59380-008
Deck-Mount Eyewash/Drench Hoses, WaterSaver Faucet
First Aid Supplies Eyewash Stations and Eyewash Bottles
Dual-head units act as a fixed eyewash or removable drench hose, providing a soft, wide stream of water that quickly and gently rinses away contaminants.
  • Supplied with a 9.5 mm (³/₈") male swivel inlet.
Feature two GS-Plus “Gentle Spray” heads angled at 45° and a brass squeeze valve with locking clip for hands-free operation. Each head has an internal filter, self-regulating flow control, and permanently attached flip-top dust cover.
Certifications: Meet ANSI Z358.1 standard.