VWR® Disposable Transfer Pipettes with Reference Lines
Supplier: VWR International
414004-036 414004-037
414004-036CS 247.72 CAD
414004-036 414004-037
VWR® Disposable Transfer Pipettes with Reference Lines
Pipettes Pasteur/Transfer Pipettes
Pipettes are designed with a small tip to ensure consistent reproduction of the drop size.
  • Polyethylene construction
  • One-piece design
These one-piece polyethylene transfer pipettes feature reference lines at 1 and 2 ml.
Ordering information: Nonsterile pipettes are bulk packaged in two easy-to-use dispenser boxes of 500 each. Sterile pipettes are individually wrapped.
Order Now
SPECIFICATIONS
Environmentally Preferable