Eyesaver® Combination Faucet and Eyewash Systems, Speakman®
Supplier: Speakman
SEF-1000-CA SEF-1800-CA-8 SEF-1800-CA-SL SEF-1801 SEF-1801-8
14224-240EA 3613.05 CAD
14224-240 97000-974 97000-978 10068-228 10068-230
Eyesaver® Combination Faucet and Eyewash Systems, Speakman®
First Aid Supplies Eyewash Stations and Eyewash Bottles
These unique eyewash systems combine a high-quality gooseneck faucet with an independently operating eyewash to provide a safer, faster lab safety system.
  • No need to activate the faucet for the eyewash to work
They are equipped with two separate supply tubes inside the gooseneck spout, one dedicated to the eyewash and the other dedicated to the faucet. This provides an independent cold or tepid water supply to the eyewash, so there is never a danger of scalding the eyes with hot water. The eyewash features aerated sprays, one-pull activation and alignment, and integral flip-top dust covers that automatically pop off when eyewash is activated. The fully functional gooseneck faucet features ¹/₄-turn ceramic valves (on wrist blade handle systems) or ceramic cartridge (on lever handle systems), configuration, and a polished chrome-plated finish.
Certifications: Meet ANSI Z358.1 standard.