Wall-Mount Eyewashes, WaterSaver Faucet
Supplier: WaterSaver Faucet
EW814 CTEW814
59380-026EA 914.98 CAD
59380-026 VFCTEW814
Wall-Mount Eyewashes, WaterSaver Faucet
First Aid Supplies Eyewash Stations and Eyewash Bottles
These wall-mounted units feature two GS-Plus 'Gentle Spray' heads to quickly and gently rinse contaminants away.
  • Heavy-duty pipe and fittings resist corrosion
The low-density polyethylene filter removes scale, deposits, and debris from the water and water line, keeping impurities out of the user’s eyes. Factory-tested and assembled units include a 29.2 cm (11¹/₂") stainless steel bowl, 1.3 cm (¹/₂") brass stay-open ball valve, pipe, fittings, and ANSI-compliant identification sign.
Certifications: Meet ANSI Z358.1 standard.