Eyesaver® Eyewash Stations, Bar Activation, Speakman®
Supplier: Speakman
SE-505 SE-545 SE-565
VF-0505-SEEA 3220.81 CAD
VF-0505-SE VF-0545-SE VF-0565-SE
Eyesaver® Eyewash Stations, Bar Activation, Speakman®
First Aid Supplies Eyewash Stations and Eyewash Bottles
These eyewash stations offer twin aerated sprays for gentle, yet thorough rinsing of the eyes and face.
  • Available in deck-mount, wall-mount, and pedestal-mount styles
The units are supplied with a rectangular stainless steel bowl and panic bar activating system.
Certifications: Meet ANSI Z358.1 standard.
Order Now
SPECIFICATIONS
Environmentally Preferable