Enclosed Base Worktables, Advance Tabco®
Supplier: Advance Tabco
CK-SS-304 CB-SS-305 CB-SS-304 CK-SS-305
58300-050EA 20004.55 CAD
58300-050 58300-056 58300-048 58300-054
Enclosed Base Worktables, Advance Tabco®
Furniture Tables and Workbenches
Enclosed base worktables with sliding doors are constructed with 14-gauge, Type 304 stainless steel tops and 18-gauge, Type 304 stainless steel bodies.
  • Tables have a 2.5 cm (1") adjustable metal bullet feet
  • Standard working height is 90 cm (35½")
Tops have a 4.1 cm (1⁵/₈") sanitary-rolled rim on front edge. Sides and rear edges are flat and turned down on a 90° angle for alignment with walls or other equipment.