Disposable Milk Pipettes, Sterile, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
56400-11 56404-11
53384-192CS 1594.25 CAD
53384-192 53384-240
Disposable Milk Pipettes, Sterile, DWK Life Sciences
Pipettes Milk Testing Pipettes
Disposable, plugged, polystyrene pipette with uniform pulled tip.
Calibrated for the measurement of undiluted and diluted milk, as prescribed in 'Standard Methods for Analysis of Dairy Products,' American Public Health Association.
Packaging: Supplied sterile in bags of 25, individually wrapped.