Disposable Milk Pipettes, Glass, Sterile, Plugged, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
72106-11 72106-22
48754-104CS 1472.49 CAD
48754-104 48754-110
Disposable Milk Pipettes, Glass, Sterile, Plugged, Kimble Chase, DWK Life Sciences
Pipettes have a uniform pulled tip and are clean, plugged, sterile, and ready for use.
The 1.1 ml size is also suitable for use in the milk phosphatase test.
Certifications: Calibrated to meet APHA requirements for making milk dilutions.
Ordering information: Packaged in paper/plastic individual wrap and overpacked in convenient dispensing shelf-packs for laboratory use.