Disposable PCR Micropipettes, Drummond
Supplier: Drummond
5-000-1001-X 5-000-1001-X10
53440-425PK 193.16 CAD
53440-425 53440-426
Disposable PCR Micropipettes, Drummond
Pipettes Microcapillaries
These disposable positive displacement micropipettes feature stainless steel plungers.
  • Graduated in 1 µl increments
  • Positive displacement micropipettes
  • Stainless steel plungers provided
  • True sample volumes measured regardless of viscosity
  • Eliminate contamination from residual aerosols
  • Decontamination at temperatures of 204 °C (400 °F) or higher