KIMAX® Reusable Measuring (Mohr) Pipettes, Class A, Color-Coded, Kimble Chase, DWK Life Sciences
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
37025-1110 37025-11100 37025-2 37025-5 37025-10 37025-25
89003-452CS 810.64 CAD
89003-452 89003-454 89003-456 89003-458 89003-451 89003-469
KIMAX® Reusable Measuring (Mohr) Pipettes, Class A, Color-Coded, Kimble Chase, DWK Life Sciences
Pipettes Measuring (Mohr) Pipettes
Calibrated to deliver, these KIMAX-51® pipettes are intended for chemical laboratory work.
  • Color coding facilitates sorting and selection
  • Designed from ASTM Specification E1293, Style 1, Class A requirements
Tip openings are smaller than typically desired for clinical laboratory applications. Scale is permanent brown stain fused into uniform bore tubing without etching. Pipettes are graduated to a base line which is on the straight tube above the taper.
Delivery information: Each pipette is supplied with a permanently marked individual serial number and a Certificate of Graduation Accuracy.