KIMAX® Reusable Measuring (Mohr) Pipettes, Class B, Color-Coded, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
37020-10 37020-1110 37020-11100 37020-2 37020-50 37020-25 37020-5 37020-110
89003-506CS 519.51 CAD
89003-506 89003-508 89003-510 89003-512 53103-191 89003-514 89003-516 72700-027
KIMAX® Reusable Measuring (Mohr) Pipettes, Class B, Color-Coded, Kimble Chase, DWK Life Sciences
Pipettes Measuring (Mohr) Pipettes
Calibrated to deliver, these rugged KIMAX-51® pipettes are intended for chemical laboratory work.
  • Color coding facilitates sorting and selection
  • Designed from ASTM Specification E1293, Style 1, Class B requirements
Tip openings are smaller than typically desired for clinical laboratory applications. Scale is permanent brown stain fused into uniform bore tubing without etching. Pipettes are graduated to a base line which is on the straight tube above the taper.
Delivery information: Each pipette is supplied with a permanently marked individualMore Product Information