KIMAX® Reusable Volumetric Pipettes, Class A, Color-Coded, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
37002-1 37006-510 37006-1 37006-1510 37006-2 37002-510 37002-1 37002-1510 37002-2 37006-2510 37006-3 37006-4 37006-5 37006-6 37006-7 37006-8 37006-9 37002-2510 37002-3 37002-4 37002-5 37002-6 37002-7 37002-8 37002-9 37002-10 37002-12 37002-15 37002-20 37002-25 37002-30 37002-40 37002-50 37002-75 37002-100 37006-10 37006-12 37006-15 37006-20 37006-25 37006-30 37006-40 37006-50 37006-75 37006-100
89002-766CS 346.78 CAD
89002-766 89003-340 89003-348 89085-708 89085-722 75856-762 75856-764 75856-766 75856-768 75856-788 75856-790 75856-792 75856-794 75856-798 75856-800 75856-802 75856-804 75856-806 75856-808 75856-810 75856-812 75856-814 75856-816 75856-818 75856-820 75856-822 75856-824 75856-826 75856-828 75856-830 75856-832 75856-834 75856-836 75856-838 75856-840 75856-842 75856-844 75856-846 75856-848 75856-852 75856-854 75856-856 75856-858 75856-860 75856-862 75856-864 75856-866 75856-868 75856-870
KIMAX® Reusable Volumetric Pipettes, Class A, Color-Coded, Kimble Chase
Pipettes Graduated Pipettes
Pipettes feature large capacity-indicating numerals on the bulb.
  • Color-coded for ease in sorting and selection
  • With a legend of permanent brown stain
  • Calibrated to deliver
Designed from ASTM Specification E969, Class A requirements. Made in compliance with ASTM Specification E1273 for color coding.
Delivery information: Serialized pipettes are supplied with a certificate of graduation accuracy.