SP Bel-Art Roto-Flo™ Flow Indicators, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H19937-0002 H19937-0003 H19937-0001 H19937-0001
47727-232CS 608.3 CAD
47727-232 47727-234 47727-238 490014-410
SP Bel-Art Roto-Flo™ Flow Indicators, Bel-Art Products, a part of SP
Flow Indicators
These indicators provide immediate visualization of flow in a tubing line.
  • Brightly colored paddle wheels spin when gas or liquid flows through a system
  • Molded of clear polystyrene
  • Smooth connecting barbs for easy installation
  • Maximum recommended pressure is 10 psig
Internal flow channel diameter is 5 mm (7/32"), paddle wheel shell is 25 mm (1"), and overall length is 51 mm (2").
Not for use with corrosive chemicals or fluids with temperatures in excess of 64 °C (150 °F).