Reusable Volumetric Pipettes, Class A, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
37003-7 37003-8 37003-9 37003-10 37003-15 37003-20 37003-25 37003-5 37003-1 37003-2 37003-3 37003-4 37003-6
89085-730CS 509.55 CAD
89085-730 75856-748 75856-750 75856-752 75856-754 75856-756 75856-758 75856-760 75856-796 75856-850 75856-872 75856-874 75856-876 75856-878
Reusable Volumetric Pipettes, Class A, Kimble Chase
Pipettes Graduated Pipettes
Calibrated to contain (bottom line) and to deliver (top line) to Class A tolerances.
The TC (to contain) feature of this pipette accommodates use with viscous liquids where the entire contents are washed out. All pipettes feature a legend of permanent brown stain and are marked with large numerals on the bulb indicating capacity. Color-coding (ASTM E1273) facilitates sorting and selection.