Airscan® MD-8 Fixed Sampling System for Microbial Air Monitoring of Controlled Environments
Supplier: Sartorius
16746-01--COM 16746-01--SHB 16746-01--SHT 16746SHBCOM 16746SHTCOM
CA76409-208EA 9507.25 CAD
CA76409-208 CA76409-212 CA76409-214 CA76409-216 CA76409-218
Airscan® MD-8 Fixed Sampling System for Microbial Air Monitoring of Controlled Environments
Samplers Air Samplers
The MD8 Airscan®, together with gelatin membrane filter disposables, constitutes a sampling system for accurate, reproducible and quantitative detection of airborne microorganisms in clean rooms and isolators, whether conventionally ventilated or under laminar flow. This system is capable of detecting even the smallest airborne contaminants, including viruses.
  • Proprietary, ready-to-use, USP-approved gelatin membrane filters
  • Compact, modular design
  • Delayed start feature
  • Stainless steel sampling head
  • Storage of calibration and test data
  • Compatibility with
More Product Information