Polystyrene Weighing Boats
Supplier: Sartorius
YBW01K YBW01L YBW01M YBW01S
CA76412-878EA 87.38 CAD
CA76412-878 CA76412-880 CA76412-882 CA76412-884
Polystyrene Weighing Boats
Boats Weighing Boats
Weighing boats, reusable, made up of anti-static polystyrene.
  • Available in small, medium, large and assorted (10 each of small, medium and large)
  • For use with CPA balances, Cubis™ balances, extend ED balances, M-power balances, M-prove scales