Med Dimensions® Canine Brachycephalic Intubation Trainer
Supplier: MED DIMENSIONS, LLC
MD-E-C1031S-1 MD-E-C1031S-2 MD-E-C1031S
470357-246EA 373.01 CAD
470357-246 470357-290 470357-230
Med Dimensions® Canine Brachycephalic Intubation Trainer
Educational Materials Biology Educational Materials Animal Biology Learning Activites
Invest in your veterinary expertise.
  • Realistic anatomy; derived from Ct scans, with relative anatomy including nasal cavity, epiglottis, and pharynx
  • Obstructed airway simulation; realistic soft palate obstruction challenges airway access, mimicking real-world difficulties
  • Replaceable soft palate is secured by magnetic clip for extended training
  • Replaceable jaw and tongue components
  • Versatile practice; manipulate with fingers, a tube, or laryngoscope
  • Ideal for use with 4 Et tube
  • Easy setup includes suction cup for mounting and threaded to fit any standard tripod
The Med Dimensions Standard Canine Brachycephalic Intubation Trainer is an affordable, portable solution for mastering this essential veterinary skill. Master a critical skill and develop the expertise and confidence to safely and effectively intubate brachycephalic patients, a notoriously challenging procedure. Enhance patient safety by reducing the risk of complications associated with difficult airways by honing your intubation technique. Optimize practice efficiency by improving procedural speed and accuracy, leading to smoother patient care and increased practice productivity. Enjoy cost-effective training that provides high-quality, hands-on training without the expense and ethical concerns of live animal practice. Benefit from flexibility and convenience by practicing at your own pace, anywhere, anytime with this portable and user-friendly trainer.
Caution: This product is a non-medical device intended solely for educational purposes. It is not designed, manufactured, or intended for use in the diagnosis, treatment, cure, mitigation, or prevention of disease or for any other medical purpose.