Microsart® e.motion Membrane Filters
Supplier: Sartorius
15406Z47SCM 28006Z47SCM
CA76517-720PK 642.05 CAD
CA76517-720 CA76517-976
Microsart® e.motion Membrane Filters
Filters Membrane Filters
Microsart® e.motion membrane filters for colony counting, particle testing and microscopy.
  • Application area: Bioburden/microbial enumeration
The membrane filters, specifically designed for use in the Microsart® e.motion dispenser, are individually sterile-sealed in a format packaged on a pleated band, without protective paper.
The special pleating of the band of membrane filters ensures that they are perfectly flat when dispensed. The membrane filters can also be used with Sartorius nutrient pad sets.
Order Now
SPECIFICATIONS
Environmentally Preferable