Thermal Transfer Cryogenic Labels with SimPEEL
Supplier: GA INTERNATIONAL INC
807657640753 807657640760 807657640876 807657640883
77321-296RL 115.42 CAD
77321-296 77321-298 77321-300 77321-302
Thermal Transfer Cryogenic Labels with SimPEEL
Labels
Thermal transfer cryo labels provide an all-in-one solution for cryo storage that can firmly adhere to ambient and already frozen surfaces, without the need for thawing, cure time, or any other special treatment.
  • Temperature range: −196 to 121 °C (−321 to 250 °F)
  • Ultra-resistant adhesive ensures no flagging or detachment when stored in liquid nitrogen
  • SimPEEL technology allows for effortless peeling and glove-friendly application
  • Thermal transfer printable for the best printout and barcode quality
  • Ideal for labeling ambient temperature as well as frozen surfaces without thawing first
  • No overlap or wrap-around required for proper adhesion
These cryogenic labels provide an all-in-one solution for cryo storage that can firmly adhere to ambient and already frozen surfaces (as low as −80 °C/−112 °F) without the need for thawing, cure time, or any other special treatment. With the addition of SimPEEL technology, these labels can now be peeled from their liner quickly and effortlessly via the adhesive-free tab that ensures the labels will not stick to hands or gloves during application and will maintain excellent long-term adhesion when stored under cryogenic conditions.
Due to their unique adhesive and conformable design, these cryo labels for frozen surfaces can be applied onto both flat and curved surfaces, ideal for identifying cryo vials, microtubes, as well as cardboard and polycarbonate storage boxes destined for long-term storage in liquid nitrogen tanks (−196 °C/−321 °F), ultra-low temperature freezers (−80 °C) and dry ice transportation. Ideally suited for identifying vials destined for immediate immersion in liquid nitrogen as well as labeling already frozen vials without having to thaw them first, thereby preserving the biological integrity of the frozen specimens.
These permanent cryo labels will also resist elevated temperatures up to 121 °C/249 °F, perfectly suitable for identifying containers destined for heated water bath thawing directly after removal from ultra-low temperature storage. Furthermore, these durable barcode labels resist standard steam autoclave sterilization conditions (121 °C/249 °F, 16psi, 20 minutes). Compatible with most common thermal transfer printers (RR-Class resin ribbon recommended for optimal printout resistance). They can also be inscribed with cryo markers.