Wrap-Around Cryogenic and Autoclave-Resistant Labels
Supplier: GA INTERNATIONAL INC
807656027647 807656017785 807656027890 807656027906 807656017792 807656017808 807656017815 807656017822
76274-052RL 235.47 CAD
76274-052 76274-054 76274-056 76274-058 76274-060 76274-062 76274-064 76274-066
Wrap-Around Cryogenic and Autoclave-Resistant Labels
Labels
Cryogenic and autoclave-resistant wrap-around labels for thermal transfer printers. These labels will withstand prolonged storage under cryogenic conditions, including in liquid nitrogen, as well as the sterilization process in steam autoclaves, dry heat ovens, and gamma irradiation.
  • Temperature range: −196 to +150 °C (−321 to +302 °F)
  • Will withstand long-term storage in cryogenic environments (−196 °C)
  • Will withstand sterilization by steam autoclaves, dry heat, and gamma irradiation
  • Lamination protects the labels against abrasion and chemical exposure
  • Thermal transfer printable with all major barcode printers
Self-laminating, autoclave-resistant labels designed for secure identification of samples in cryogenic storage. The wrap-around transparent part of the label securely laminates the printable part of the label and protects against exposure to chemicals and physical damage. Wrap-around labels can withstand temperatures up to 150 °C.