Direct Thermal Cryogenic Labels with SimPEEL
Supplier: GA INTERNATIONAL INC
807657631584 807657631591
77321-106RL 76.41 CAD
77321-106 77321-108
Direct Thermal Cryogenic Labels with SimPEEL
Labels
Direct thermal printable cryogenic labels for already frozen surfaces. The unique SimPEEL technology makes these cryo labels easy to peel and apply, even on frozen surfaces.
  • Temperature range: −196 to +70 °C (−321 to +158 °F)
  • Quick labeling of frozen vials and tubes (no thawing required)
  • Withstands exposure to solvents, water baths, and condensation
  • Compatible with most common direct thermal printers
  • Convenient to print, no need for ribbons or other ink sources
These solvent-resistant cryogenic direct thermal printable labels can easily attach to already frozen vials, frozen surfaces, and frozen containers (as low as −80 °C/−112 °F). Labeling samples without having to thaw them first makes these labels an ideal choice for identifying valuable temperature-sensitive samples and specimens. Moreover, these direct thermal labels have been enhanced with the unique patent pending SimPEEL™ technology, allowing them to be easily and quickly peeled from their liner. These permanent cryogenic labels are designed to be stable in low temperature environments, including long-term storage in liquid nitrogen (−196 °C/−321 °F), and ultra-low temperature freezers (−80 °C/−112 °F), as well as dry ice transport.
In addition, these labels display excellent resistance to routinely used laboratory chemicals like detergents, alcohols (100% ethanol immersion: up to 15 minutes), xylene (immersion: up to 30 minutes), other organic solvents, disinfecting solutions, as well as surface-sanitizing wipes.
These labels are ideal for use in research institutes, clinical laboratories, biobanks, biotechnology companies, and other facilities that store samples under low temperature conditions.