TruSpan® Universal Face Shield
Supplier: Hexarmor
TruSpan®
17-36001
77591-766EA 100.65 CAD
77591-766
TruSpan® Universal Face Shield
Face Shields
Preassembled and ready for action, TruSpan® adapts to multiple competitor helmet models and to deliver comfortable, close-fitting face protection, complete with permanent TruShield® anti-fog and scratch-resistant technology.
  • Pre-assembled and ready for immediate use
  • System allows tight-nested locking and resting positions for less strain and more balance
  • Added sealing lip where TruSpan® meets the helmet’s brim protects against dirt/debris and chemical/splash risks
  • Rubber headgear band is designed with robust, long-lasting EPDM material
  • Components are dielectric, making system suitable for electrical work
  • Wide coverage around the face with panoramic view
  • Meets D3 liquid splash protection + 100% UV protection
When not in use, TruSpan® has tight nesting capabilities on top of the helmet for more compact, balanced comfort, or it can be easily removed. TruSpan® can be used with select slot-adapted earmuffs for a fully integrated system, as the helmets’ side slots are not needed for attachment.
Compatible with but not limited to: Ceros® XP300; Bullard C30, S62, C33; Honeywell Fiber Metal E1, Fiber Metal E2, North Zone, North A89; 3M H-700, G3001; MSA V-Gard full brim, V-Gard cap style, V-Gard 500
Compatible with select slot-adapted earmuffs, as headgear does not interfere with helmets’ side slots for attachment.
Easily detach face shield when in nested position with a simple squeeze and pull of the side arms without removing head gear from helmet.
Face shield includes permanent TruShield® anti-fog coating on the inside and scratch-resistant hard coat on the outside.
Certifications: ANSI/ISEA Z87.1-2020 (Impact Protection Z87+, Liquid Splash Protection D3); CSA Z94.3:20; EN 166:2001; CE.