VWR® Maximum Protection Zipper Labcoats
Supplier: VWR International
76336-676 76336-678 76336-680 76336-854 76338-314 76336-677 76336-679 76336-681 76336-855 76338-315 76532-280 76532-282 76532-284 76532-286 76532-288 76532-290 76532-292
76336-676CS 6450.23 CAD
76336-676 76336-678 76336-680 76336-854 76338-314 76336-677 76336-679 76336-681 76336-855 76338-315 76532-280 76532-282 76532-284 76532-286 76532-288 76532-290 76532-292
VWR® Maximum Protection Zipper Labcoats
Disposable Garments
VWR® Maximum Protection zipper lab coats are engineered for durability and comfort, making them an essential choice for maintaining compliance and safety in critical environments.
  • Full zipper front closure ensures a secure fit and easy donning/doffing while maintaining maximum protection
  • Durable fabric made from high-quality materials provides excellent protection against particulates, chemicals, and liquids
  • Elastic wrists provide a snug fit to prevent contaminants from entering the sleeve area (also available with knit cuffs)
  • Knit collar adds comfort while maintaining a professional appearance and secure barrier
  • Lightweight and breathable design for comfort during extended wear in controlled environments
  • Low-lint fabric minimizes particle shedding
  • Crucial for maintaining cleanroom integrity
VWR® Maximum Protection Zipper lab coats are specifically designed for environments where safety, cleanliness, and protection are paramount. These lab coats provide superior barrier protection against contaminants, making them ideal for use in cleanrooms, laboratories, pharmaceutical manufacturing, and biosafety level facilities. The provide a reliable barrier against particulate contamination, ensuring cleanliness and sterility during sensitive production processes. Ideal for high-level containment facilities where maximum protection is critical, these garments offer protection from chemical splashes and biological hazards.
Ideal for use in ISO Class 5 - 8 cleanrooms to ensure operations remain compliant with cleanliness standards. The material construction combines comfort with durability for long wear periods without sacrificing protection. The zipper closure allows quick and secure fitting, improving efficiency in high-paced environments. All styles feature one chest pocket and two hip pockets. These lab coats are available with knit cuffs or elastic wrists.