IPTG (isopropyl-β-D-thiogalactopyranoside) ≥98% (by HPLC), high purity animal-free, Millipore®

Supplier: MilliporeSigma

420291-1GM 420291-5GM
CA95061-828EA 313.82 CAD
CA95061-828 CA95061-830
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥98% (by HPLC), high purity animal-free, Millipore®
IPTG (Isopropyl-β-D-thiogalactopyranoside)
Formula: C₉H₁₈O₅S
Storage Temperature: Refrigerator
MDL Number: MFCD00063273
CAS Number: 367-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR