Centrifuge Tubes with Conical Skirted Base
Supplier: SARSTEDT INC
58.618 58.618.008 58.619
77587-694CS 198.95 CAD
77587-694 77587-696 77587-698
Centrifuge Tubes with Conical Skirted Base
Tubes Centrifuge Tubes
High-quality, safe and robust containers are essential for mixing, processing and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, Sarstedt offers the right product for every laboratory and analytical requirement.
  • Base shape: skirted conical base
  • Tube with skirted base ensures that it stands securely
  • Polystyrene (PS) tubes for maximum transparency
  • Polypropylene (PP) tubes for thermal, mechanical and chemical applications
  • Different push caps for a variety of applications
This broad product portfolio in the field of reagent and centrifuge tubes includes options with round, conical or flat base; with or without skirted base; with stopper or screw cap; with or without print or labels.